Learn More
MilliporeSigma™ Supelco™ SPE Tube Adapter configured for AutoTrace Automated Systems
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods.
Specifications
Packaging Quantity 6/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
111020922
View Documents Promotion Details
MilliporeSigma™ Supelco™
57123
3 mL Tubes N/A
Only null left
111012129
View Documents Promotion Details
MilliporeSigma™ Supelco™
57126
6 mL Tubes N/A
Only null left
Description
Description
Specifications
Specifications
6/Pk.