missing translation for 'onlineSavingsMsg'
Learn More

Solstice SPE Frit

Catalog No. 6002985
Change view
Click to view available options
For Use With (Equipment):
1.5 mL Reservoir
25 mL Reservoir
4.0 mL Reservoir
75 mL Reservoir
8.0 mL Reservoir
Quantity:
10,000
100
1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
6002985 75 mL Reservoir 100
6002986 1.5 mL Reservoir 100
6002987 4.0 mL Reservoir 100
6003028 8.0 mL Reservoir 100
6003130 25 mL Reservoir 10,000
6003131 25 mL Reservoir 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 6002985 Supplier Solstice Supplier No. 9444

For Reservoir

  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality

Specifications

For Use With (Equipment) 75 mL Reservoir
Quantity 100