Learn More
Solstice SPE Columns, THC (C8 + Aminopropyl), 500 mg/3 mL
Catalog No. 6003120
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
6003120 50/Pk.
1 options
Catalog No. 6003120 Supplier Solstice Supplier No. 9914
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
Specifications
Quantity 50/Pk.