Learn More
Solstice SPE Columns, Drug Abuse (C8 + Benzyl Sulfonic Acid), 500 mg/3 mL
Catalog No. 6003117
Change view
Click to view available options
Description:
30/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
6003117 30/Pk.
6003116 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6003117 Supplier Solstice Supplier No. 9906
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical-grade polypropylene with PE frits,
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
Specifications
Description 30/Pk.