missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPC Hazwik™ 20-Gallon Lab Pack Spill Kit
Designed for small to medium spills of hazardous materials
Supplier: Brady Worldwide Inc SKH20
Includes: Contents packed in high-quality 20-gal. drum with screw-top lid:
- Twelve 15 x 19 in. pads
- Three 3 in. x 12 ft. Socks
- Two 17 x 19 in. Pillows
- One pair nitrile gloves
- Three disposal bags
- One pair goggles
- Emergency Response Handbook
Description
Order Info
Weight: 26 lb. (11.8kg).
Compliance
UN approved