missing translation for 'onlineSavingsMsg'
Learn More

SPC Drum Top Covers

Catalog No. 19819441
Change view
Click to view available options
For Use With (Equipment):
55 gal. Dimpled Drums
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19819441 55 gal. Dimpled Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819441 Supplier Brady Worldwide Inc Supplier No. DTO25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Product Type Pre-cut Drum Top Cover
Color White
For Use With (Application) Absorbs only Oil, prevent pump drips from reaching the floor and keep drum tops and drum storage areas clean
For Use With (Equipment) 55 gal. Dimpled Drums
Includes 1 or 2 Bung Holes
Diameter (English) Inner 22 in.
Diameter (Metric) Inner 55.8 cm