missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Spatula
Spatula with one rounded angled end and one square end.
Supplier: Synthware™ W160150
Description
- Available in various lengths.
Specifications
| 1 Ea. | |
| Round End, Tapered End | |
| Spoon Spatula | |
| 5.9 in. | |
| 150 mm |
| Autoclavable | |
| Silver | |
| Organic Synthesis | |
| Metal |