Learn More
Moldex™ SparkPlugs™ Ecostation™ Ear Plug Dispenser
Catalog No. 19384371
Change view
Click to view available options
CSA Z94.2:
Class AL
Catalog No. CSA Z94.2
19384371 Class AL
1 options
Catalog No. 19384371 Supplier Moldex™ Supplier No. MDX6703
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SparkPlugs™ EcoStation™ dispensers pre-filled with ear plugs. Easy access, wall-mountable unit. When empty, simply dispose of it and hang a new one.
  • More economical and green than separately packaged ear plugs
  • Easy-to-install sealed refills snap in quickly
  • Empty bottles are fully recyclable
  • Gives workers easy access to hearing protection
  • 500 pairs in each EcoStation
  • High visibility to increase awareness for hearing conservation
  • Hang anywhere ear plugs are needed
  • Workers take one pair at a time with no wasteful grabbing of multiple pairs
  • Independently tested to ANSI S3.19-1974
  • 100% PVC-free
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Energy, Utilities, Manufacturing, Healthcare, Mining, Oil and Gas, Transportation, and Other Various Industries
High Visibility No
Holds 500 pairs
Packaging Type Dispenser
Pairs per Package 500 Pairs/Dispenser
Plug Material Foam
Plug Color Multi-color
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Mounting Wall Mountable
Show More Show Less