Learn More
Ted Pella Inc SPARKLE OPTICAL LENS CLNR 4OZ DFS Item
Supplier:  Ted Pella Inc 80919
A transparent non-ammonia, non-alcohol, safe cleaner for high tech lenses. apply Sparkle Optical Lens Cleaner to surface to be cleaned and wipe with clean Lens Cleaning Cloth. Excellent for microscopes, magnifiers, eye glasses and safety goggles. Do not use on contact lenses. anti-Static properties limit dust attraction. Testimonials from universities and private labs as the first non-alcohol product that is safe for objective lens use. (4oz)
Catalog No. NC0090079
Only null left
Explore available promotions