Learn More
Brady™ Spare LED Bulb for Sign Projector, Clear
Catalog No. 17823775
Change view
Click to view available options
Wattage:
40 W
300 W
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wattage
17823775 40 W
17823776 300 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17823775 Supplier Brady™ Supplier No. 178984
Restores original projector brightness for optimal visibility.
  • Specifically designed to replace the existing LED in compatible projectors.
  • Copper mounting plate ensures efficient heat transfer for optimal LED performance and lifespan.
  • Replacement of the LED requires soldering and basic electrical skills and should be performed by a qualified electrician.
  • LED can only be operated when mounted onto the projector heat sink otherwise damage can occur.
Specifications
Product Type Bulb
Display LED
Wattage 40 W