Learn More
Agilent BioTek Spare AMX manifold
Spare AMX manifold set, contains aspirate and dispense manifolds
Supplier:  Agilent BioTek 1260036
Catalog No. BT1260036
Only null left
Description
Description
  • Includes one dispense and one aspirate manifold
  • For use with 1260035 (one each), sold separately
Specifications
Specifications
CE and TUV marked. RoHS compliant.
Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization
Spare AMX manifold set, aspirate and dispense manifolds
Spare AMX manifold set
MFXP2R
Product Suggestions
Product Suggestions