missing translation for 'onlineSavingsMsg'
Learn More

Span 83 (=Sorbitan Sesquioleate), TCI America™

Catalog No. S006325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S006325G 25 g
S0063500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S006325G Supplier TCI America Supplier No. S006325G

Chemical Identifiers

CAS 8007-43-0
MDL Number MFCD00151568
Synonym Arlacel 83, Arlacel C, Sorbitan Sesquioleate

Specifications

Color Yellow
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Span 83 (=Sorbitan Sesquioleate)
EINECSNumber (8)-0063
RTECSNumber WG2934330
TSCA Yes