missing translation for 'onlineSavingsMsg'
Learn More

Span 60 (=Sorbitan Monostearate), TCI America™

Catalog No. p-7135789
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S006225G 25 g
S0062500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S006225G Supplier TCI America Supplier No. S006225G
Only null left

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
Synonym sorbitan stearate, sorbitan stearate inn, span 60 tn, sorbitan monostearate nf
PubChem CID 23725022
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Melting Point 52°C
Color Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Span 60 (=Sorbitan Monostearate)
EINECSNumber (8)-0063
RTECSNumber WG2933500
TSCA Yes