missing translation for 'onlineSavingsMsg'
Learn More

AIMS™ Space Gel Heating Pad

Catalog No. 14370223
Encompass
Change view
Click to view available options
For Use With (Equipment):
Standard thermal plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14370223 Standard thermal plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14370223 Supplier AIMS™ Supplier No. GGP1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optional gel heating pad maintains temperature of around 33anddeg; to 34anddeg;C depending on the room temperature.

  • Contains enough material for 30 students when working in groups of 2 to 3

Specifications

For Use With (Application) Maintains temperature of around 33° to 34°C depending on room temperature
Type Space Gel Heating Pad
For Use With (Equipment) Standard thermal plate