missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SP600125, 98%

Catalog No. AAJ67272MF
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67272MF 50 mg
AAJ67272MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67272MF Supplier Thermo Scientific Chemicals Supplier No. J67272MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 129-56-6 | C14H8N2O, C14H8N2O | 220.23 g/mol

SP 600125 is a potent and reversible inhibitor of JNK1-3 with an IC50 value of 0.11 M.1 It is cell-permeable and dose-dependently inhibits c-Jun phosphorylation in cells, blocking the expression of COX-2 and TNF-α in monocytes and IL-10, TNF-α, and IFN-γ in T-cells. SP 600125 also prevents apoptosis in many cell types, including B-cells, and inhibits autophagy in HeLa cells. Competitively and reversibly inhibits JNK1, 2 and 3 (IC50 = 40 - 90 nM) with negligible activity at ERK2, p38β and a range of enzymes (IC50 > 10 ?M). Shown to have reduced selectivity over other protein kinases under certain conditions. Prevents BMP9-induced osteogenic differentiation of MSCs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129-56-6
Molecular Formula C14H8N2O
Synonym 1,9-Pyrazoloanthrone;

Specifications

Quantity 50 mg
Description Yellow to brown
Content And Storage -30°C to -10°C
Formula Weight 220.23
Solubility Soluble in DMSO at approx. 20mg/ml
Product Type SP600125, 99%
Form Solid
Assay 99%

RUO – Research Use Only