Learn More
Bel-Art™ Pipette Pump™ Pipetter
Provides safe, accurate. and trouble-free pipetting. Pipetters utilize an easy-to-use one-handed operation.
$96.95 - $96.95
Specifications
Adjustable Tip Spacing No
Disposable Reusable
For Use With Serologic Pipette
Grip Type Ergonomic
Material Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
S35821
View Documents
Bel-Art™
F378980000EMD
Green 10 mL N/A
Only null left
Description
Description
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121°C (250°F) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available
Specifications
Specifications
No
Serologic Pipette
Polypropylene
1
Pipette
Reusable
Ergonomic
Pipette Pump Pipettors
Non-sterile
Acid and Alkali
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'