Learn More
Bel-Art™ Large Pails
Rugged pails with fitted lids/covers and steel-plated bail handles.
Supplier:  Bel-Art™ H16772
 View more versions of this product
Catalog No. S30518B
Only null left
Description
Description
  • Available in yellow or translucent white for easier viewing of fluid level
  • Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
  • 14-qt./13.2L pail has steel-plated swinging bail handle with grip
  • Large pails meet US Specification MIL-P36257A
Compliance
Meet US Specification MIL-P36257A (8/10/70).
Specifications
Specifications
Pail
None
Not Autoclavable
Product Suggestions
Product Suggestions