missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners

Catalog No. 1199918
Change view
Click to view available options
Dimensions (L x W):
24 x 20 in. (610 x 508mm)
50 ft. x 20 in.
Color:
Orange
White
Description:
One single roll, 50 ft.
Pack of 100 sheets
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color Description
1199918 50 ft. x 20 in. White One single roll, 50 ft.
1199919 50 ft. x 20 in. Orange One single roll, 50 ft.
1199920 24 x 20 in. (610 x 508mm) White Pack of 100 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1199918 Supplier Bel-Art™ Supplier No. 246750000
Get Up to $500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials

Specifications

Product Type Bench Protector
Material Polyethylene
Absorbancy 400 mL/m2
Length (English) 50 ft.
Length (Metric) 15.2 m
Width (English) 20 in.
Width (Metric) 508 mm
Dimensions (L x W) 50 ft. x 20 in.
Color White
Description One single roll, 50 ft.
Disposable Single-use
For Use With (Application) Simplify cleanup and safeguard work surfaces from stains, spills and wear
Show More Show Less