missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners

Catalog No. S41267
Change view
Click to view available options
Dimensions (L x W):
50 ft. x 20 in.
Color:
White
Quantity:
50 ft./Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color Quantity
S41267 50 ft. x 20 in. White 50 ft./Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S41267 Supplier Bel-Art™ Supplier No. F24675 0000
Get Up to $500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials

Specifications

Product Type Bench Protector
Material Polyethylene
Absorbancy 420 mL/m2
Length (English) 50 ft.
Length (Metric) 15.2 m
Width (English) 20 in.
Width (Metric) 50.8 cm
Dimensions (L x W) 50 ft. x 20 in.
Color White
Disposable Single-use
For Use With (Application) Simplify cleanup and safeguard work surfaces from stains, spills and wear
Quantity 50 ft./Roll
Show More Show Less