missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ 96 Deep-Well Plates

Catalog No. 03421154
Change view
Click to view available options
Product Type:
Deepwell Plate
Sterile; 96 Deep-Well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
03421154 Sterile; 96 Deep-Well Plates
22038083 Deepwell Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03421154 Supplier Bel-Art™ Supplier No. F378600001
Get Up to $500 in Products with Purchase
View all promotions

Expanded capability for multi-channel pipetting

  • Offer the working volume of culture tubes and the convenience of a 96-well plate format.
  • Can be used for variety of processes and operations such as sequencing, precipitations and dilution.
  • Compatible with deep-freezing work or sample storage.
  • Ideal for manual 8 and 12 multi-channel pipetting procedures.
  • Compatible with robotic sampling processors and automated liquid handling systems.
  • Includes Zymark™, Tecan™, Hamilton Scientific™, Rasys™ and Beckman™.
  • Side load collars required for side loading Beckman Biomek Systems.
  • Plates are molded of autoclavable polypropylene.
  • Each 2 mL well has round bottom for easy sample retrieval, alphanumerically identified.
  • Steam autoclavable at 121°C (250°F).
  • Dimensions: 5 x 3.37 x 1.62 in. H
  • An optional, specially designed polypropylene (PP), keyed fit cover separates each well with a sealing ring to reduce evaporation and permits safe and stable stacking of plates.
  • Covers are keyed to fit only one way.

Order Info

Individually packaged and available sterile or non-sterile

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Natural
Product Type Sterile; 96 Deep-Well Plates
Material Polypropylene
Volume (Metric) Well 2 mL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
For Use With (Application) Used for variety of processes and operations such as sequencing, precipitations and dilution
Show More Show Less