missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soybean Oil, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Soybean Oil, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Grade | USP |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Soybean Oil |