missing translation for 'onlineSavingsMsg'
Learn More

Soybean oil

Catalog No. AAJ61399AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61399AP 500 mL
AAJ61399K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61399AP Supplier Thermo Scientific Chemicals Supplier No. J61399AP

CAS: 8001-22-7

Soybean oil is used as a base for printing inks and oil paints. It is also used as a fixative to extend the short duration of action of essential oils such as geranium oil in several commercial products. Further, it is used on citrus fruits and domestic dwellings. It is also employed in the determination of freezing point of soybean oil. It plays as a delivery vehicle of alfa-lipoic acid. In addition to this, it is used as a condiment for salads as well as involved in the synthesis of epoxidized-soybean oil (ESO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Soybean oil is used as a base for printing inks and oil paints. It is also used as a fixative to extend the short duration of action of essential oils such as geranium oil in several commercial products. Further, it is used on citrus fruits and domestic dwellings. It is also employed in the determination of freezing point of soybean oil. It plays as a delivery vehicle of alfa-lipoic acid. In addition to this, it is used as a condiment for salads as well as involved in the synthesis of epoxidized-soybean oil (ESO).

Solubility
Miscible with absolute alcohol, ether, petroleum ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 8001-22-7
MDL Number MFCD00132356
Synonym Glycine Soja

Specifications

Color Colorless to Yellow
Density 0.92
Flash Point >113°C (235°F)
Refractive Index 1.4743
Quantity 500 mL
Solubility Information Miscible with absolute alcohol,ether,petroleum ether. Immiscible with water.
Physical Form Oil
Chemical Name or Material Soybean oil
EINECSNumber 232-274-4
RTECSNumber WG4862000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only