missing translation for 'onlineSavingsMsg'
Learn More

PAN, 1% (w/v) Indicator Solution in Methanol, Spectrum™ Chemical

Catalog No. 18612121
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18612121 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18612121 Supplier Spectrum Chemical Mfg Cor Supplier No. P101100ML
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical indicator solution for laboratory use.

Specifications

Chemical Name or Material PAN
CAS 85-85-8, 67-56-1
Concentration 1%
Packaging Amber Glass Bottle
Color Colorless
Quantity 100 mL
Grade Chemical Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.