Learn More
SouthernBiotech™ Fluoromount-G™ Slide Mounting Medium
Catalog No. OB10001
Change view
Click to view available options
Volume (Metric):
25 mL
Catalog No. Volume (Metric)
OB10001 25 mL
1 options
Catalog No. OB10001 Supplier SouthernBiotech™ Supplier No. OB010001
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Water soluble, nonfluorescing mounting medium
  • Ideal for immunofluorescent staining procedures
  • Particularly well-suited for slide staining procedures with an aqueous final step
  • Supplied in sufficient quantity to mount approximately 500 slides
Specifications
Applications Immunofluorescent Staining Procedures
Volume (English) 0.84 oz.
Volume (Metric) 25 mL