Learn More
Thermo Scientific™ Sorvall ST4 Plus Centrifuge Series
Faster processing of more samples with a high capacity-to-footprint ratio and simple programming interface, plus the flexibility to run most tubes and bottles on one centrifuge.
Supplier:  Thermo Scientific™ 75016043FLASH
 View more versions of this product
Catalog No. 75016043PM
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Easy to Use:
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
  • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific™ Auto-Lock™ III Rotor System:
  • Secure locking system allows easy push-button installation and exchange of rotors.
  • System saves time when switching between applications.
  • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Thermo Scientific™ ClickSeal™ Bucket Sealing System:
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
  • System has glove-friendly, one-handed open/close capability.
  • System is certified by CAMR™ in Porton Down, UK.
Thermo Scientific™ SMARTSpin™ Technology:
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
  • Technology optimizes safety and improves separations.
Space-Saving Design:
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Energy Efficient:
  • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
Chemical Identifiers
EN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Bright LCD Push-Button User Interface
Less waste, Energy efficient
Up to 6 Programs via LCD User Interface buttons
Up to 9 hr. 59 min.
Option to count up or down
5 Year Limited
120 V
Microprocessor
Sorvall
Direct, Brushless Induction Low Profile
Choices of 9 Accel/10 Decel Rates
Yes
ISO 14971, ISO 13485, ISO 9001
Benchtop Centrifuge
120 V
60 Hz
Specifications
EN/UL/IEC 61010-1EN/UL/IEC 61010-2FCC part 15
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
CFC free
Direct, Brushless Induction Low Profile
Sorvall ST4R Plus-MD TX-1000 Blood Tube Package
Up to 6 Programs via LCD User Interface buttons
Up to 9 hr. 59 min.
-10°C to +40°C, ±2°C
Benchtop Centrifuge
120 V
60 Hz
Microprocessor
Sorvall
Bright LCD Push-Button User Interface
Less waste, Energy efficient
Choices of 9 Accel/10 Decel Rates
Yes
ISO 14971, ISO 13485, ISO 9001
Option to count up or down
5 Year Limited
120 V
Product Suggestions
Product Suggestions