missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Microtube Adapters

Catalog No. 76003750
Change view
Click to view available options
For Use With (Equipment):
0.2mL microtubes
For 0.25/0.4mL microtubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
76003750 0.2mL microtubes
76003759 For 0.25/0.4mL microtubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76003750 Supplier Thermo Scientific™ Supplier No. 76003750

Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ adapters.

Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.

Ordering Information:

Sold in packs of 24

Specifications

For Use With (Equipment) 0.2mL microtubes
Color Gray
No. per Pack 24