missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall BIOS 16 Adapters

Adapters (set of 2 adapters) for Thermo Scientific Sorvall BIOS 16 centrifuges

Specifications

Type Adapter
No. per Pack 2
View More Specs

Products 6
Catalog Number Mfr. No. Tube Type No. of Places Price Quantity & Availability  
Catalog Number Mfr. No. Tube Type No. of Places Price Quantity & Availability  
75003341
View Documents Promotion Details
Thermo Scientific™
75003341
5/7 mL Blood Collection Tube 52 N/A
Only null left
 
75003342
View Documents Promotion Details
Thermo Scientific™
75003342
Blood Collection 42 N/A
Only null left
 
75003343
View Documents Promotion Details
Thermo Scientific™
75003343
Conical 26 N/A
Only null left
 
75003344
View Documents Promotion Details
Thermo Scientific™
75003344
Conical 11 N/A
Only null left
 
75003345
View Documents Promotion Details
Thermo Scientific™
75003345
Standard Microplate or Deepwell Plate 5 Microplates or 1 DW Plate N/A
Only null left
 
75003346
View Documents Promotion Details
Thermo Scientific™
75003346
500 mL Conical Bottle 1 N/A
Only null left
 
Description

Description

Adapters dedicated to Thermo Scientific Sorvall BIOS 16 centrifuges, dedicated to large volume and bioprocessing applications. These adapters fit into the 2000 mL buckets of the 6x2000 mL rotor (Part Number 75003962) and 2000 mL buckets of the 8x2000 mL rotor (Part number 75003982) equipped with 2000 mL buckets (Part number 75003964). They are available as set of 2 adapters.

Thermo Scientific Sorvall BIOS 16 centrifuges include features that not only help enhance reliability, but make them much simpler to use. Whether you require outstanding capacity or high performance for application flexibility, they offer features that will help maximize productivity—and when combined with our space-saving footprint, one of these options is sure to be a perfect fit in your bioprocessing facility. These adapters fit into the 2000 mL buckets of the 6x2000 mL rotor (Part Number 75003962) and 2000 mL buckets of the 8x2000 mL rotor (Part number 75003982) equipped with 2000 mL buckets (Part number 75003964). These adapters are available as set of 2 adapters.
Specifications

Specifications

Adapter
2
Videos
SDS
Documents

Documents