Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
Catalog No. 75007621
Change view
Click to view available options
Tube Type:
15 mL DIN
7 mL Blood
7 mL DIN
Conical
Microtube
Tube Size:
1.5/2 mL
7 mL
15 mL
50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tube Type Tube Size
75007621 Conical 15 mL
13100592 Conical 50 mL
13100595 15 mL DIN 15 mL
13100600 Microtube 1.5/2 mL
13100599 7 mL Blood 7 mL
13100598 7 mL DIN 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 75007621 Supplier Thermo Scientific™ Supplier No. 75007621
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Tube Type Conical
For Use With (Equipment) 4-place Swing rotor with round bucket
Tube Size 15 mL