Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Specifications
| Type | Adapter |
| Tube Type | Conical |
| Capacity | 50 mL |
| For Use With (Equipment) | 4-place Swing rotor with round bucket |
| Tube Size | 50 mL |