missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan sesquioleate

An emulsifier | CAS: 8007-43-0 | C42H76O7

Supplier:  Thermo Scientific Chemicals J62648K2

 View more versions of this product

Catalog No. AAJ62648K2


Only null left
Explore available promotions

Description

Description

Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel 'coated' after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel ‘coated‘ after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

Solubility
Insoluble in cold water, hot water. soluble in mineral oil and ethyl acetate.

Notes
Keep container tightly closed. Keep container in a cool, well-ventilated area and protect from moisture. 
Specifications

Specifications

Sorbitan sesquioleate
8007-43-0
0.989
113°C (235°F)
C42H76O7
Arlacel™ 83; Span™ 83
693.06
Yellow
Viscous Liquid
1 L
1.478
MFCD00151568
Insoluble in cold water,hot water. soluble in mineral oil and ethyl acetate.

Chemical Identifiers

8007-43-0
C42H76O7
MFCD00151568
Arlacel™ 83; Span™ 83
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 232-360-1

RTECSNumber : WG2934330

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only