missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan sesquioleate

Catalog No. AAJ62648AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62648AP 500 mL
AAJ62648K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62648AP Supplier Thermo Scientific Chemicals Supplier No. J62648AP

An emulsifier | CAS: 8007-43-0 | C42H76O7

Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel 'coated' after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel ‘coated‘ after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

Solubility
Insoluble in cold water, hot water. soluble in mineral oil and ethyl acetate.

Notes
Keep container tightly closed. Keep container in a cool, well-ventilated area and protect from moisture. 

Chemical Identifiers

CAS 8007-43-0
Molecular Formula C42H76O7
MDL Number MFCD00151568
Synonym Arlacel™ 83; Span™ 83

Specifications

Color Yellow
Physical Form Viscous Liquid
Density 0.989
Quantity 500 mL
Flash Point 113°C (235°F)
Refractive Index 1.478
Solubility Information Insoluble in cold water,hot water. soluble in mineral oil and ethyl acetate.
Formula Weight 693.06
Chemical Name or Material Sorbitan sesquioleate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 232-360-1
RTECSNumber WG2934330
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only