Learn More
Sorbitan monostearate
CAS: 1338-41-6 | C24H46O6 | 430.63 g/mol
Supplier: Thermo Scientific Chemicals L114350B
| Quantity | 1000 g |
|---|
Description
Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.
Solubility
Soluble in ethanol (50 mg/ml), isopropanol, mineral oil and vegetable oil. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
| C24H46O6 | |
| MFCD00005366,MFCD00005366 | |
| Sorbitan monooctadecanoate; Span™ 60 | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
Specifications
| Sorbitan monostearate | |
| 53°C to 57°C | |
| 1000 g | |
| C24H46O6 | |
| 14,8724 | |
| Soluble in ethanol (50mg/ml),isopropanol,mineral oil and vegetable oil. Insoluble in water. | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O | |
| 430.63 | |
| 430.63 |
| 1338-41-6 | |
| 1 | |
| >149°C (300°F) | |
| MFCD00005366,MFCD00005366 | |
| Sorbitan monooctadecanoate; Span™ 60 | |
| HVUMOYIDDBPOLL-IIZJTUPISA-N | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate | |
| 23725022 |
Safety and Handling
EINECSNumber : 215-664-9
RTECSNumber : WG2933500
TSCA : Yes
Recommended Storage : Ambient temperatures