missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitan Monostearate, FCC, 27-34%, Spectrum™ Chemical
S1318, 1338-41-6, C24H46O6
Supplier: Spectrum Chemical Mfg Cor S131825KG
Description
Spectrum™ Chemical Sorbitan Monostearate, FCC is used as an emulsifier to keep oils and water mixed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 1338-41-6 | |
| Poly Pail | |
| MFCD00005366,MFCD00005366 | |
| 147-157 | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O | |
| 430.63 | |
| FCC |
| 1 | |
| C24H46O6 | |
| 2.5 kg | |
| HVUMOYIDDBPOLL-IIZJTUPISA-N | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate | |
| 27 to 34% |
Chemical Identifiers
| 1338-41-6 | |
| 430.63 | |
| HVUMOYIDDBPOLL-IIZJTUPISA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
| C24H46O6 | |
| MFCD00005366,MFCD00005366 | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate |