missing translation for 'onlineSavingsMsg'
Learn More

Sorafenib tosylate

Catalog No. AC465712500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465712500 250 mg
AC465710010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465712500 Supplier Thermo Scientific Chemicals Supplier No. 465712500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 475207-59-1 | C28H24ClF3N4O6S | 637.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 475207-59-1
Molecular Formula C28H24ClF3N4O6S
Molecular Weight (g/mol) 637.03
InChI Key IVDHYUQIDRJSTI-UHFFFAOYSA-N
IUPAC Name 4-[4-({[4-chloro-3-(trifluoromethyl)phenyl]carbamoyl}amino)phenoxy]-N-methylpyridine-2-carboxamide; 4-methylbenzene-1-sulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.CNC(=O)C1=CC(OC2=CC=C(NC(=O)NC3=CC=C(Cl)C(=C3)C(F)(F)F)C=C2)=CC=N1

Specifications

Quantity 250 mg
Solubility Information Solubility aq. soln.: < 1 g/L (25°C). Other solubilities: < 1 g/L ethanol (25°C), 127 g/L dmso
Formula Weight 637.03
Chemical Name or Material Sorafenib tosylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only