missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Sonometer Wire

Catalog No. S23832
Change view
Click to view available options
Length (Metric):
1.5 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S23832 1.5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23832 Supplier Eisco™ Supplier No. PH0730A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With ends looped ready for use.

  • Set of six wires of brass and steel
  • Ends looped ready for use
  • Available in lengths of 1.5m of 22, 24, 26 SWG

Specifications

Product Type Sonometer Wire
For Use With (Application) Available in lengths of 1.5m of 22, 24, 26 SWG
Includes Set of six wires of brass and steel
Length (Metric) 1.5 m