Learn More
Sonoco™ ThermoSafe Foam-Only Dome Style EPS Insulated Shippers
EPS foam-only shippers offer solution for safe storage and delivery of most refrigerated or frozen products
Supplier: Sonoco™ TS333
Description
- Large selection of cost-effective, reusable stock containers available
- Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter
Specifications
| 8/Cs. | |
| Insulated Shipper | |
| 37.5 x 33.6 x 25.7 cm (14.87 x 13.25 x 10.12 in.) | |
| Expanded Polystyrene Insulated |