missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sonoco™ ThermoSafe EPS Foam and Corrugate Shippers
Large selection of cost-effective, reusable EPS foam in corrugate stock containers available
Supplier: Sonoco™ 05REFOT
Description
- EPS foam containers in 200 lb. test corrugated carton
- Excellent dry ice sublimation… average dry ice usage in 1-1/2 inch thick wall units is 5 pounds over 24 hours and 3-4 pounds thereafter
- Standard 1-1/2 inch wall shippers are ideal for shipments of 24 to 48 hours
Specifications
| 15.5 in. | |
| Rounded Corners and Edges | |
| 9 in. | |
| 6 in. | |
| 12.5 in. | |
| 91kg Test Corrugated Carton | |
| 0.5 kg |
| 15.5 in. | |
| Safe storage and delivery of refrigerated or frozen material (pharmaceuticals, laboratory specimens, serums, vaccines, human organs, culture pack and nuclear and isotope materials) | |
| 12.5 in. | |
| 15.24 cm | |
| 1 lb. | |
| <24 hr. | |
| Molded Expanded Polystyrene |