Learn More
Sonoco™ ThermoSafe DiagnosticShipper Systems, Without Tubes
Ready-to-use combination/triple packaging kits
Supplier:  Sonoco™ 571
 View more versions of this product
Catalog No. 0352557
Only null left
Description
Description
Tested and certified to meet the material and performance requirements for the transport of diagnostic specimens in conformance with U.S. Department of Transportation (DOT), U.S. Postal Service and International Air Transport Association (IATA) Packing Instruction 650.
Specifications
Chemical Identifiers
Conforms with DOT, USPS, IATA packing instruction 650
Diagnostic specimen shipping system without tubes
Specifications
2 envelope, absorbents, bubble wrap
Conforms with DOT, USPS, IATA packing instruction 650
Diagnostic specimen shipping system without tubes
Product Suggestions
Product Suggestions