Learn More
SOMNI Scientific™ SOMNI Scientific™ Rebreathing Bag
The bag is designed to inflate uniformly and has a reinforced neck. Latex, PVC and BPA free
Supplier:  SOMNI Scientific™ BB12012
 View more versions of this product
Catalog No. 04777202
Only null left
Description
Description
  • The rebreathing bag is designed to inflate uniformly and has a reinforced neck
  • Latex free
  • PVC and BPA free
Specifications
Chemical Identifiers
Research Animal Inhalant Anesthesia
Breathing Bag
Specifications
Research Animal Inhalant Anesthesia
Breathing Bag
2 L
Product Suggestions
Product Suggestions