Learn More
SOMNI Scientific™ SOMNI Scientific™ CO2 H-Tank Reg
Description
- Attaches to a large h size tank and supplies a constant flow of carbon dioxide up to 12 LPM
- Gas specific regulator for CO2
- H-tank
- Flows from 0-12 LPM
Specifications
Specifications
| For Use With (Application) | Research Animal Inhalant Anesthesia |
| Includes | Flowmeter |