missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Solvent-Resistant White Biopsy Bags

Catalog No. 6774022
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
6774022 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6774022 Supplier Epredia™ Supplier No. 6774022

Solvent-resistant nylon and thin mesh provide, and allow for optimal fluid exchange around the tissue

  • Minimize the risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Rapidly remove specimens with convenient peel-open bag

Specifications

Product Type Biopsy Bag
Material Nylon, Solvent-resistant
Color White
Length (Metric) 31 mm
Width (Metric) 43 mm
Length (English) 1.2 in.
Width (English) 1.7 in.
Dimensions (L x W) 1.2 x 1.7 in.
Size Small
Quantity 1000/Cs.