missing translation for 'onlineSavingsMsg'
Learn More

Solvent Black 5, TCI America™

Catalog No. O009725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O009725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O009725G Supplier TCI America Supplier No. O009725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solvent dye used for staining in microbiology and research applications.

Chemical Identifiers

CAS 11099-03-9
Molecular Formula C8H19ClN2O
Molecular Weight (g/mol) 194.703
MDL Number MFCD00071316
InChI Key GBPWTMZRCAESKB-UHFFFAOYSA-N
PubChem CID 19828004
IUPAC Name 2-amino-N,N-dipropylacetamide;hydrochloride
SMILES CCCN(CCC)C(=O)CN.Cl

Specifications

Color Black
Quantity 25 g
Physical Form Crystal/Powder
Grade Research
Chemical Name or Material Solvent Black 5
EINECSNumber (5)-3142
TSCA Yes