Learn More
Perkin Elmer Canada Solution-E6100DRC Sensitivity/Det Limit
Catalog No. 509045610
Encompass
Part of the Solution Kit which includes items: 2 x 1,000 mL: Setup/Stability/Masscal Solution, 2 x 250 mL: Wash Solution, 1 x 250 mL: Sensitivity/Detection Limit Solution, 1 x 125 mL: Methanol Blank Solution, 1 x 125 mL: Chromium in Methanol Solution.
Catalog No. 509045610 Supplier Perkin Elmer Canada Supplier No. N8125034
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Part of the Solution Kit which includes items: 2 x 1,000 mL: Setup/Stability/Masscal Solution, 2 x 250 mL: Wash Solution, 1 x 250 mL: Sensitivity/Detection Limit Solution, 1 x 125 mL: Methanol Blank Solution, 1 x 125 mL: Chromium in Methanol Solution.