missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Solution dNTPs: PCR Nucleotide Mix
Description
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Specifications
| Format | Liquid |
| Product Type | PCR Nucleotide Mix dNTP Mix (10mM each A,C,G,T) |
| Concentration | 10 mM |
| Purity | >99% |
| Quantity | 500 μL |
| Color | Colorless |
| For Use With (Application) | Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |