Description
Solution D is an anion transport inhibitor, for cells such as CHO-K1, that can be used in conjunction with CCF2-AM and CCF4-AM-based studies to prevent active cellular export of the FRET-based substrates after loading.Specifications
| Pharma & Biopharma; Protein Assays and Analysis; Cellular Imaging | |
| GeneBLAzer | |
| 1 mL |
For Research Use Only. Not for use in diagnostic procedures.