Learn More
Solution D
Catalog No. K1157
Encompass
Change view
Click to view available options
Quantity:
1 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1157 25 mL
K1156 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K1157 Supplier Thermo Fisher Scientific Supplier No. K1157
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Solution D
Solution D is an anion transport inhibitor, for cells such as CHO-K1, that can be used in conjunction with CCF2-AM and CCF4-AM-based studies to prevent active cellular export of the FRET-based substrates after loading.
Spécifications
For Use With (Application) Pharma & Biopharma; Protein Assays and Analysis; Cellular Imaging
Product Line GeneBLAzer
Quantity 25 mL
For Research Use Only. Not for use in diagnostic procedures.