Learn More
3M™ Solus™ CCS Series Eyewear
Catalog No. 19071106
Change view
Click to view available options
Lens Tint:
Clear
Includes:
Solus Eyewear
Catalog No. Lens Tint Includes
19071106 Clear Solus Eyewear
1 options
Catalog No. 19071106 Supplier 3M™ Supplier No. 7100196396
Only null left
3M™ Solus™ CCS Series eyewear includes the Corded Control System (CCS) temples. The versatile design helps keep eyewear and earplugs attached, untangled and ready to use, for convenient, comfortable protection needed to focus on the jobs at hand. Earplugs sold separately.
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings. 
  • Optional: Comfortable gasket that helps limit eye exposure to debris while providing additional cushioning
  • Polycarbonate lens absorbs 99.9% UVA & UVB and meet the requirements of ANSI Z87.1- 2015 Standard as an Impact Rated spectacle
  • 3M Catalog# SCCS01SGAF-GRN, SCCS02SGAF-GRN, SCCS02SGAF-GRN-F, SCCS07SGAF-GRN, SCCS07SGAF-GRN-F
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Certifications/Compliance (Europe) ANSI Z87.1-2015
Lens Tint Clear
Product Type Protective Eyewear
For Use With (Application) Masonary, Food and Beverage, Aviation, Primary Metals, Chemical Handling
Frame Color Green
Includes Solus Eyewear
No. per Case 20/Cs.
Temple Style Cord Control System Temples
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless
Show More Show Less