missing translation for 'onlineSavingsMsg'
Learn More

MAPA Professional Solo 980 Nitrile Disposable Gloves

Catalog No. 17467380
Change view
Click to view available options
Size:
2X-Large, 10
3X-Large, 11
Large, 8
Medium, 7
Small, 6
X-Large, 9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17467380 Small, 6
17467381 Medium, 7
17467382 Large, 8
17467383 X-Large, 9
17467378 2X-Large, 10
17467379 3X-Large, 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17467380 Supplier MAPA Professional Supplier No. 34980456

Disposable nitrile glove providing enhanced mechanical and chemical protection for demanding tasks.

  • Thick nitrile construction delivers excellent mechanical resistance and long-lasting durability in tough environments.
  • Certified high-level PPE ensures dependable protection against hazardous substances and biological risks.
  • Approved for food contact applications, making it suitable for food handling and processing tasks.
  • Latex-free and powder-free design reduces allergy risk and enhances wearer comfort during extended use.
  • Pebbled fingertips improve grip and control, even in oily or slippery environments.
  • Ambidextrous and versatile, ideal for industrial, laboratory, and chemical-handling applications.
  • Complies with Regulation (EC) No. 1272/2008 and REACH Regulation (EC) No. 1907/2006, free from hazardous classification, substances of very high concern (SVHC), and annex XVII restricted substances.
  • UE type certificate / CE type examination certificate: 0075/014/162/10/24/2331.

Specifications

Accelerator Free No
Applications Laboratory, Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 8 mil
Finger Thickness (Metric) 0.2 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 8 mil
Palm Thickness (Metric) 0.2 mm
Powdered Powder Free
Product Type Nitrile Gloves
Size Small, 6
Sterility Non-sterile
Thickness (English) 8 mil
Thickness (Metric) 0.2 mm
Glove Cuff Style Rolled
Model SOLO 980
Series SOLO 980
Length (English) 12 in.
Length (Metric) 30 cm
Product Line SOLO 980
Show More Show Less