Learn More
Eisco™ Solid Stainless Steel Hooked Weights
Solid stainless steel hooked weights with integrated hooks. Weights are cylindrical with natural stainless steel color, weight stamped at top
Supplier:  Eisco™ PHHWSS3
 View more versions of this product
Catalog No. S41604
Only null left
Description
Description
  • Solid stainless steel construction
  • Hook is integrated with weight
  • Natural stainless steel color with stamped identifier at top
  • Weight is cylindrical
Warranty
1 year limited manufacturer warranty
Specifications
Chemical Identifiers
Cylindrical Weight
Calibrating scales and balances
1
Stainless Steel
Weight with hook
Specifications
Cylindrical Weight
50 g
Calibrating scales and balances
1
Stainless Steel
2.54 cm
Weight with hook
Product Suggestions
Product Suggestions