missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, T-bore PTFE Stopcock

Solid Phase Peptide Synthesis Vessel with a side arm and a coarse or medium porosity fritted glass disc.
$411.25 - $537.86
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass |
| Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
31501574
![]() |
Synthware™
P130010C |
10 mL | Vessel, Coarse Porosity |
Each for $411.25
|
|
||||
31501575
![]() |
Synthware™
P130050C |
50 mL | Vessel, Coarse Porosity |
Each for $495.61
|
|
||||
31501576
![]() |
Synthware™
P130050M |
50 mL | Vessel, Medium Porosity |
Each for $495.61
|
|
||||
31501577
![]() |
Synthware™
P130100C |
100 mL | Vessel, Coarse Porosity |
Each for $537.86
|
|
||||
31501578
![]() |
Synthware™
P130100M |
100 mL | Vessel, Medium Porosity |
Each for $537.86
|
|
||||
Specifications
| Organic Synthesis | |
| Borosilicate Glass |